Learn More
Aviva Systems Biology Corp GSTZ1 Antibody - N-terminal region (ARP49036_T100) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49036_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GSTZ1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GSTZ1. Alternate names: GSTZ1-1, MAAI, MAI, MGC2029ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60001