Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSTZ1 Mouse anti-Human, Clone: OTI6A10, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527332

Catalog No. PIMA527332


Add to cart

Description

Description

This gene is a member of the glutathione S-transferase (GSTs) super-family which encodes multifunctional enzymes important in the detoxification of electrophilic molecules, including carcinogens, mutagens, and several therapeutic drugs, by conjugation with glutathione. This enzyme also plays a significant role in the catabolism of phenylalanine and tyrosine. Thus defects in this enzyme may lead to severe metabolic disorders including alkaptonuria, phenylketonuria and tyrosinaemia. Several transcript variants of this gene encode multiple protein isoforms.
Specifications

Specifications

GSTZ1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O43708
GSTZ1-1, MAAI, MAI, S-(hydroxyalkyl)glutathione lyase, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, glutathione S-aryltransferase, glutathione s-transferase Zeta 1, maleylacetoacetate isomerase, maleylacetone isomerase
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2954
Western Blot
OTI6A10
Unconjugated
GSTZ1
Liquid
GSTZ1
Full length human recombit protein of GSTZ1 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit