Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GSX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144269
Description
GSX1 Polyclonal Antibody for IHC (P), ELISA
GSX1 gene ontology annotations related to this gene include adenohypophysis development; hypothalamus development; neuron fate commitment; spinal cord association neuron differentiation.
Specifications
GSX1 | |
Polyclonal | |
Unconjugated | |
GSX1 | |
genomic screened homeo box 1; GS homeo box protein 1; GS homeobox 1; Gsh1; Gsh-1; GSX1; Homeobox protein GSH-1 | |
Rabbit | |
Protein G | |
RUO | |
219409 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
4.92 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q9H4S2 | |
GSX1 | |
Recombinant Human GS homeobox 1 protein (67-147AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction