Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GT 2016, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
High affinity H3 receptor antagonist (Ki = 43.8 nM). Displays selectivity against H1 and H2 receptors (IC50 >10 μM). Increases the release of histamine in the cerebral cortex. Displays no activity at histamine methyltransferase in vitro at concentrations up to 3 μM. Brain penetrant.
Specifications
Quantity | 50 mg |
Formula Weight | 317.47 |
Percent Purity | >98% |
Chemical Name or Material | GT 2016 |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction