missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GT 2016, Tocris Bioscience™
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
Description
High affinity H3 receptor antagonist (Ki = 43.8 nM). Displays selectivity against H1 and H2 receptors (IC50 >10 μM). Increases the release of histamine in the cerebral cortex. Displays no activity at histamine methyltransferase in vitro at concentrations up to 3 μM. Brain penetrant.
Specifications
Quantity | 10 mg |
Formula Weight | 317.47 |
Percent Purity | >98% |
Chemical Name or Material | GT 2016 |
Safety and Handling
Recommended Storage | Store at -20°C |
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'