Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTDC1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$482.50

Specifications

Antigen GTDC1
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123431
SDP
View Documents
Novus Biologicals
NBP309265100UL
100 μg
Each of 1 for $482.50
Add to Cart
 
Description

Description

GTDC1 Polyclonal specifically detects GTDC1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

GTDC1
Western Blot
Unconjugated
Rabbit
Human
FLJ11753, FLJ44822, glycosyltransferase-like 1, glycosyltransferase-like domain containing 1, glycosyltransferase-like domain-containing protein 1, mannosyltransferase candidate, mat-Xa
The immunogen is a synthetic peptide directed towards the N-terminal region of human GTDC1 (XP_011510164.1). Peptide sequence LIIEAFYGGSHKQLVDLLQEELGDCVVYTLPAKKWHWRARTSALYFSQTI
Affinity purified
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
PBS buffer, 2% sucrose
79712
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit