Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTDC1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15307820UL

 View more versions of this product

Catalog No. NBP15307820


Add to cart

Description

Description

GTDC1 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

GTDC1
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
GTDC1
Synthetic peptides corresponding to GTDC1(glycosyltransferase-like domain containing 1) The peptide sequence was selected from the N terminal of GTDC1. Peptide sequence CQERDFQYGYNQILSCLVADVVVFNSVFNMESFLTSMGKFMKLIPDHRPK.
Immunogen affinity purified
RUO
Primary
79712
Western Blot
Western Blot 1:100-1:2000
Q4AE62
FLJ11753, FLJ44822, glycosyltransferase-like 1, glycosyltransferase-like domain containing 1, glycosyltransferase-like domain-containing protein 1, mannosyltransferase candidate, mat-Xa
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit