Learn More
Aviva Systems Biology Corp GTF2A1 ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp P100987_P050
Encompass_Preferred
Rabbit polyclonal antibody recognizes the C terminal of GTF2A1 (general transcription factor IIA, 1, 19/37kDa) protein. It is suitable for WB and IHC applications; 50ug/50ul
Catalog No. 50-014-62817
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.