Learn More
Aviva Systems Biology Corp GTF2B Antibody - N-terminal region (P100989_P050) | 100uL
Supplier:  Aviva Systems Biology Corp P100989_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GTF2B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GTF2B. Alternate names: TF2B, TFIIBApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Zebrafish
Catalog No. 50-014-62818