Learn More
Aviva Systems Biology Corp GTF2E1 Antibody - C-terminal region (P100892_P050) | 100uL
Supplier:  Aviva Systems Biology Corp P100892_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GTF2E1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human GTF2E1. Alternate names: FE, TF2E1, TFIIE-AApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Mouse, Rabbit, Rat
Catalog No. 50-014-62768