Learn More
Aviva Systems Biology Corp GTF2F1 Antibody - N-terminal region (P100810_P050) | 100uL
Supplier:  Aviva Systems Biology Corp P100810_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GTF2F1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GTF2F1. Alternate names: BTF4, RAP74, TF2F1, TFIIFApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rabbit, Zebrafish
Catalog No. 50-014-62720