Learn More
GTF2F2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 151391AP
Catalog No. 50-173-0014
Add to cart
Description
Description
GTF2F2 is required for both basal and HIV-1 Tat-activated transcription. GTF2F2 synergizes with HIV-1 Tat and the cellular co-activator Tat-SF1 during Tat-mediated transactivation of the HIV-1 LTR promoter.
Specifications
Specifications
GTF2F2
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P13984, Q01750, Q8R0A0
ATP dependent helicase GTF2F2, BTF4, GTF2F2, RAP30, TF2F2, TFIIF, TFIIF beta
Rabbit
IgG
150 μL
-20°C
Primary
2963, 68705, 81674
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
GTF2F2
Liquid
Gtf2f2
GTF2F2 Fusion Protein Ag7313
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat