Learn More
Aviva Systems Biology Corp GTF2H1 Antibody - N-terminal region (P100811_T100) | 100uL
Supplier:  Aviva Systems Biology Corp P100811_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GTF2H1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human GTF2H1. Alternate names: BTF2, TFB1, TFIIHApplicationIHC, WB. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-62721