Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF2H1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

$395.30

Specifications

Antigen GTF2H1
Concentration 0.5 mg/mL
Applications Western Blot
Conjugate Unconjugated
Format Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-172-6080
Encompass_Preferred
View Documents
Proteintech Group IncSupplier Diversity Partner
101731AP150UL
150 μL
Each for $395.30
Add to cart
 
Description

Description

TFIIH, a multisubunit complex is involved in several biological fundamental mechanisms of the cell: transcription, nucleotide excision repair and cell cycle regulation. p62 is one of the six subunits that constitutes the core of TFIIH. Analysis of the expression of the p62 gene reveals an over expression in testis tissue. This subunit of TFIIH participates in a variety of protein interactions. For example, Rb competes with TBP and p62 for binding to E2F thus repressing E2F mediated transactivation; herpes simplex virus VP16 and human p53, directly interact with the p62 subunit of TFIIH. In addition, TFIIH, via p62 phosphorylation is the major target for mitotic inactivation of transcription. GTF2H1 is a component of the core GTF2H basal transcription factor involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II. GTF2H1 is one of the six subunits forming the core GTF2H basal transcription factor.
Specifications

Specifications

GTF2H1
Western Blot
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Polyclonal
-20°C
0.5 mg/mL
Unconjugated
Rabbit
Human, Mouse
IgG
Antigen Affinity Chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit