Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF2H2 Mouse anti-Human, Clone: PCRP-GTF2H2-1B9, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen GTF2H2
Clone PCRP-GTF2H2-1B9
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Applications Flow Cytometry, Immunofluorescence, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122170
View Documents
Novus Biologicals
NBP30734820UG
20 μg
Each for $138.37
Add to cart
 
NB122169
View Documents
Novus Biologicals
NBP30734810UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

GTF2H2 Monoclonal antibody specifically detects GTF2H2 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Protein Array
Specifications

Specifications

GTF2H2
0.2 mg/ml
Flow Cytometry, Immunofluorescence, Peptide Array
Purified
RUO
Human
Basic transcription factor 2 44 kDa subunit, BTF2, BTF2 p44, BTF2P44TFIIH basal transcription factor complex p44 subunit, General transcription factor IIH polypeptide 2, general transcription factor IIH subunit 2, general transcription factor IIH, polypeptide 2 (44kD subunit), general transcription factor IIH, polypeptide 2, 44kD subunit, general transcription factor IIH, polypeptide 2, 44kDa, MGC102806, T-BTF2P44, TFIIH
Recombinant full-length human GTF2H2 protein
Monoclonal
Protein A or G purified
PCRP-GTF2H2-1B9
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Stem Cell Markers
10 mM PBS with 0.05% BSA
2966
IgG1
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit