Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GTF2H4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567338
Description
Immunogen sequence: EFSKAQEESTG LLSGLRIWHT QLLPGGLQGL ILNPIFRQNL RIALLGGGKA WSDDTSQLGP DKHARDVPSL DKYAEERWEV VL Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.
GTF2H4 belongs to the TFB2 family. It is a component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II.
Specifications
GTF2H4 | |
Polyclonal | |
Unconjugated | |
GTF2H4 | |
AW545633; basic transcription factor 2 52 kDa subunit; BTF2 p52; general transcription factor II H, polypeptide 4; general transcription factor IIH polypeptide 4; General transcription factor IIH subunit 4; general transcription factor IIH, polypeptide 4, 52kDa; GTF2H4; p52; TFB2; TFIIH; TFIIH basal transcription factor complex p52 subunit; TFIIH transcription/DNA repair factor p52 subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2968 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q92759 | |
GTF2H4 | |
Recombinant Human GTF2H4. Recombinant protein control fragment (Product #RP-107987). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction