Learn More
Aviva Systems Biology Corp GTPBP1 Antibody - N-terminal region (ARP51625_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51625_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GTPBP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GTPBP1. Alternate names: GP-1, GP1, HSPC018, MGC20069ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60572