Learn More
GTPBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group Inc 163741AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0902
Add to cart
Description
Description
Promotes degradation of target mRNA species. Plays a role in the regulation of circadian mRNA stability. Binds GTP and has GTPase activity.
Specifications
Specifications
GTPBP1
Polyclonal
Unconjugated
GTPBP1
G protein 1, GP 1, GP1, GTP binding protein 1, GTPBP1, HSPC018
Rabbit
Antigen Affinity Chromatography
RUO
14904, 300077, 9567
-20°C
Liquid
Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
D2XV59, O00178, O08582
GTPBP1
GTPBP1 Fusion Protein Ag9332
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG