Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTPBP9 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157453
Description
GTPBP9 Polyclonal specifically detects GTPBP9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GTPBP9 | |
Polyclonal | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Q9NTK5 | |
OLA1 | |
Synthetic peptides corresponding to GTPBP9 (GTP-binding protein 9 (putative)) The peptide sequence was selected from the N terminal of GTPBP9. Peptide sequence MIGPIIDKLEKVAVRGGDKKLKPEYDIMCKVKSWVIDQKKPVRFYHDWND. | |
100 μL | |
Proteases & Other Enzymes | |
29789 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
DKFZp313H1942, DNA damage-regulated overexpressed in cancer 45 protein, DOC45, EC 3.6.3, EC 3.6.3.-, GBP45, GTP-binding protein 9, GTP-binding protein 9 (putative), GTP-binding protein PTD004, GTPBP9, homologous yeast-44.2 protein, Obg-like ATPase 1, PTD004 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Rat: 100%; Human: 100%; Xenopus: 100%; Western clawed frog: 100%; Chicken: 100%; Sumatran orangutan: 100%; Green puffer: 100%; Bovine: 100%; Mouse: 100%; Atlantic salmon: 92%; Rainbow smelt: 92%; Zebrafish: 92%; Fruit fly: 85%; Argulus sp. Arg2: 85%; Southern house mosquito: 78%; Pea aphid: 78%; Hanseniella sp. 'Han2': 78%; Orchesella imitari: 78%; Tomocerus sp. 'Tom2': 78%; Savannah tsetse fly: 78%; African malaria mosquito: 78%; Giant whipscorpion: 78%;. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title