Learn More
Invitrogen™ GTSE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113749
Description
Immunogen sequence: IDLMTNTPDM NKNVAKPSPV VGQLIDLSSP LIQLSPEADK ENVDSPLLKF For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 86%; Dog: 100%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 93%; Rat: 100%.
The protein encoded by this gene is only expressed in the S and G2 phases of the cell cycle, where it colocalizes with cytoplasmic tubulin and microtubules. In response to DNA damage, the encoded protein accumulates in the nucleus and binds the tumor suppressor protein p53, shuttling it out of the nucleus and repressing its ability to induce apoptosis.
Specifications
GTSE1 | |
Polyclonal | |
Unconjugated | |
GTSE1 | |
B99; G two S phase expressed protein 1; G2 and S phase-expressed protein 1; G-2 and S-phase expressed 1; G2 and S-phase expressed 1; GTSE1; GTSE-1; Protein B99; Protein B99 homolog; RGD1563164 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51512 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NYZ3 | |
GTSE1 | |
Synthetic peptide directed towards the middle region of human GTSE1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.