Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanabenz Acetate, USP, 98-101.5%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
Packaging:
Amber Glass Bottle
23256-50-0
C10H12Cl2N4O2
291.13
MCSPBPXATWBACD-GAYQJXMFSA-N
N''-[(E)-[(2,6-dichlorophenyl)methylidene]amino]guanidine; acetic acid
CC(O)=O.NC(N)=N\N=C\C1=C(Cl)C=CC=C1Cl
23256-50-0
C10H12Cl2N4O2
291.13
MCSPBPXATWBACD-GAYQJXMFSA-N
N''-[(E)-[(2,6-dichlorophenyl)methylidene]amino]guanidine; acetic acid
CC(O)=O.NC(N)=N\N=C\C1=C(Cl)C=CC=C1Cl

G1391, 23256-50-0, C8H8Cl2N4•C2H4O2

Spectrum™ Guanabenz Acetate, USP is an alpha agonist and used as an antihypertensive drug. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.002
Quantity 500 g
Loss on Drying 0.01
CAS Max % 1
UN Number UN2811
Percent Purity 98 to 101.5%