Guanfacine hydrochloride, Tocris

α2A agonist

Manufacturer: R&D Systems 1030/10

Quantity 10mg

 View more versions of this product

Catalog No. 10-301-0

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C9H9Cl2N3O·HCl
    Formula Weight 282.56
    Synonym [(2, 6-Dichlorophenyl)acetyl]guanidine hydrochloride
    Chemical Name or Material Guanfacine hydrochloride

    Specifications

    Formula Weight 282.56
    Quantity 10mg
    Assay Percent Range >99%
    Molecular Formula C9H9Cl2N3O·HCl
    CAS 29110-48-3
    Synonym [(2, 6-Dichlorophenyl)acetyl]guanidine hydrochloride
    Recommended Storage Store at Room Temperature
    Chemical Name or Material Guanfacine hydrochloride

    Store at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit