Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine Carbonate, 99+%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
593-85-1
0·5 H2CO3
90.08
MFCD00013029
593-85-1
0·5 H2CO3
90.08
MFCD00013029
Melting Point 267.0°C to 270.0°C
Color White
Infrared Spectrum Authentic
Quantity 1kg
Packaging Plastic bottle
Assay Percent Range 99+%
Linear Formula [C(NH2)3]2CO3
Beilstein 03,82
Solubility Information Solubility in water: 450g/L (20 c). Other solubilities: slightly soluble in alcohol and acetone
Formula Weight 90.08
Physical Form Crystalline Powder
Percent Purity ≥99%
Chemical Name or Material Guanidine carbonate, 99+%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 209-813-7

RTECSNumber : FG1750000

TSCA : TSCA