Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2), ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
10kg
25kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum
50-01-1
CH5N3·HCl
95.53
PJJJBBJSCAKJQF-UHFFFAOYSA-N
guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride
5742
CHEBI:32735
guanidine;hydrochloride
C(=N)(N)N.Cl
50-01-1
CH5N3·HCl
95.53
PJJJBBJSCAKJQF-UHFFFAOYSA-N
guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride
5742
CHEBI:32735
guanidine;hydrochloride
C(=N)(N)N.Cl
Specific Gravity 1.34
Melting Point 179.0°C to 186.0°C
Quantity 250g
Water 0.3% max.
Color White to Yellow
Density 1.3400g/mL
Infrared Spectrum Authentic
Packaging Plastic bottle
Assay Percent Range 98%
Linear Formula H2NC(=NH)NH2·HCl
Beilstein 03,82
Merck Index 15,4597
Solubility Information Solubility in water: 2280g/L (20°C). Other solubilities: 300g/L ethanol (30°C), 19.5g/100g dimethylformamide (30°C), <0.05g/100g acetone -<0.05g/100g benzene (30°C), insoluble in trichloroethylene
Formula Weight 95.53
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Guanidine hydrochloride
Show More Show Less

GHS P Statement
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning