Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Guanidine hydrochloride, 98%

Click to view available options
Quantity:
250g
1000g
5000g
50-01-1
CH6ClN3
95.53
MFCD00013026
PJJJBBJSCAKJQF-UHFFFAOYSA-N
guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride
5742
CHEBI:32735
guanidine;hydrochloride
C(=N)(N)N.Cl
50-01-1
CH6ClN3
95.53
MFCD00013026
PJJJBBJSCAKJQF-UHFFFAOYSA-N
guanidine hydrochloride,guanidinium chloride,guanidine, monohydrochloride,aminoformamidine hydrochloride,aminomethanamidine hydrochloride,guanidine monohydrochloride,guanidine hcl,guanidine chloride,iminourea hydrochloride,carbamidine hydrochloride
5742
CHEBI:32735
guanidine;hydrochloride
C(=N)(N)N.Cl

Alfa Aesar Guanidine hydrochloride, 98%, Quantity: 1000g, Melting Point: 178 deg.C to 185 deg.C, Molecular Weight: 95.53, Percent Purity: 98%, Beilstein: 3591990, CAS: 50-01-1, ChEBI: CHEBI:32735, Density: 1.344, EINECS Number: 200-002-3, Formula Weight: 95.53

Alfa Aesar Guanidine hydrochloride, 98%, Quantity: 1000g, Melting Point: 178 deg.C to 185 deg.C, Molecular Weight: 95.53, Percent Purity: 98%, Beilstein: 3591990, CAS: 50-01-1, ChEBI: CHEBI:32735, Density: 1.344, EINECS Number: 200-002-3, Formula Weight: 95.53

Applications
Guanidine hydrochloride is used for purification of proteins and nucleic acids. It can be used as a medicine, organic synthesis intermediate and is used in dyes. It acts as an intermediate in the preparation of sulfadiazine, which is an important raw material for sulfamethyldiazine, sulfamethazine and folic acid. It is utilized in the synthesis of 2-amino-pyrimidine, 2- amino -6- methyl-pyrimidine and 2-amino -4,6 - dimethyl-pyrimidine. Also used in RNA isolation to dissociate nucleoproteins and inhibit RNase.

Solubility
Soluble in water and ethanol.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.May agglomerate upon storage.
Quantity 250g
Sensitivity Hygroscopic
Density 1.344
Melting Point 178°C to 185°C
Beilstein 3591990
Merck Index 144562
Solubility Information Soluble in water and ethanol.
Formula Weight 95.53
Percent Purity 98%
Chemical Name or Material Guanidine hydrochloride

Causes serious eye irritation, May form combustible dust concentrations in air, Causes skin irritation, Harmful if swallowed or if inhaled

Serious eye damage/eye irritation (category 2), Skin corrosion/irritation (category 2), Acute toxicity (category 4)

EINECSNumber : 200-002-3

RTECSNumber : MF4300000

TSCA : Yes

Recommended Storage : Ambient temperatures

RMF - Research Use or Further Mfg