Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Guanine hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Guanine hydrochloride is one of the strongest denaturant. It is mainly used in protein purification and protein folding studies. It is a nucleobase used in biochemical research.
Solubility
Slightly soluble in water
Notes
Store in a cool place. Hygroscopic. Incompatible with strong oxidizing agents.
Guanine hydrochloride is one of the strongest denaturant. It is mainly used in protein purification and protein folding studies. It is a nucleobase used in biochemical research.
Solubility
Slightly soluble in water
Notes
Store in a cool place. Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Melting Point | >300°C |
Assay Percent Range | 99% |
Quantity | 10 g |
Beilstein | 4159901 |
Merck Index | 14,4564 |
Solubility Information | Slightly soluble in water |
Formula Weight | 187.59 |
Percent Purity | 99% |
Chemical Name or Material | Guanine hydrochloride |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement H315-H319-H335 |
Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a |
EINECSNumber | 211-235-5 |
RTECSNumber | MF8400000 |
TSCA | Yes |
Recommended Storage | Keep cold |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction