Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Guanosine-5'-Triphosphate Disodium Salt, Thermo Scientific™

Click to view available options
Quantity:
25mg
100mg
500mg
1g
Packaging:
Plastic bottle
56001-37-7
C10H14N5Na2O14P3
567.144
MFCD00083629
SKAJZVYOHNIUFB-LGVAUZIVSA-N
guanosine 5'-triphosphate,guanosine-5'-triphosphate disodium salt dihydrate gtp,guanosine-triphosphate,guanosine 5'-triphosphate 4-,gtp 4-,2r,3s,4r,5r-5-2-amino-6-hydroxy-9h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxy phosphonatooxy phosphinate,2r,3s,4r,5r-5-2-amino-6-oxo-3h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl phosphate
131676145
disodium;(2R,3R,4S,5R)-2-(2-amino-6-oxo-3H-purin-9-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate
C1=NC2=C(N1C3C(C(C(O3)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)[O-])[O-])NC(=NC2=O)N.[Na+].[Na+]
56001-37-7
C10H14N5Na2O14P3
567.144
MFCD00083629
SKAJZVYOHNIUFB-LGVAUZIVSA-N
guanosine 5'-triphosphate,guanosine-5'-triphosphate disodium salt dihydrate gtp,guanosine-triphosphate,guanosine 5'-triphosphate 4-,gtp 4-,2r,3s,4r,5r-5-2-amino-6-hydroxy-9h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxy phosphonatooxy phosphinate,2r,3s,4r,5r-5-2-amino-6-oxo-3h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl phosphate
131676145
disodium;(2R,3R,4S,5R)-2-(2-amino-6-oxo-3H-purin-9-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate
C1=NC2=C(N1C3C(C(C(O3)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)[O-])[O-])NC(=NC2=O)N.[Na+].[Na+]
Formula Weight 567.14
Physical Form Crystalline Powder
Packaging Plastic bottle
Quantity 25mg
Chemical Name or Material Guanosine-5'-Triphosphate Disodium Salt