Alfa Aesar™ Guanosine, 98+%

Manufacturer: Alfa Aesar™ A1132814

Quantity 25g

 View more versions of this product

Catalog No. AAA1132814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 118-00-3
    Molecular Formula C10H13N5O5
    Formula Weight 283.24
    MDL Number MFCD00010182
    Beilstein 625911
    Merck Index 14,4566
    Chemical Name or Material Guanosine

    Specifications

    Formula Weight 283.24
    CAS 118-00-3
    Beilstein 625911
    Merck Index 14,4566
    Optical Rotation −72° (c=1.5 in 0.1N NaOH)
    RTECS Number MF8750000
    TSCA TSCA
    Molecular Formula C10H13N5O5
    MDL Number MFCD00010182
    Melting Point ≈250°C (dec.)
    Quantity 25g
    Chemical Name or Material Guanosine
    Assay ≥98%

    Description

    Applications
    Guanosine is used as a constituent of nucleic acids. It is used in metallic paints, simulated pearls,plastics,cosmetics industry etc,. It has been also used in pharmacokinetics as a prodrug. Guanosine is used in cell culture applications as a precursor of GMP.

    Solubility
    Soluble in Formic Acid:Water (1:1): 50 mg/mL. Insoluble in methanol, diethyl ether.

    Notes
    Store away from extreme heat. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit