Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Guanylate kinase Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297222PCP
Description
Guanylate kinase Polyclonal antibody specifically detects Guanylate kinase in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
Guanylate kinase | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
EC 2.7.4.8, FLJ42686, FLJ43710, GMK, GMP kinase, guanylate kinase, guanylate kinase 1 | |
Produced in rabbits immunized with E. coli-derived Human Guanylate kinase fragment. | |
0.1 mL | |
Cancer, Signal Transduction | |
2987 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction