Learn More
Guanylyl Cyclase alpha 1 Mouse anti-Human, Allophycocyanin, Clone: 3G6B2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237401APC
Catalog No. NBP237401T
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Guanylyl Cyclase alpha 1
3G6B2
Flow Cytometry
Purified
GUCY1A3
Purified recombinant fragment of human GUCY1A3 (AA: 22-214) expressed in E. Coli.
Protein A or G purified
Signal Transduction, Vision
Primary
2982
Flow Cytometry
APC
PBS with 0.05% Sodium Azide
EC 4.6.1.2, GC-S-alpha-1, GCS-alpha-1, GCS-alpha-3, guanylate cyclase 1, soluble, alpha 3, guanylate cyclase soluble subunit alpha-3, GUC1A3GUCA3, GUCSA3GC-SA3, GUCY1A1, Soluble guanylate cyclase large subunit
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human