Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guar Gum, FCC, 70%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
9000-30-0
C10H14N5Na2O12P3
535.15
JEKDCIBJADJZSK-UHFFFAOYNA-L
disodium hydrogen [({[5-(6-amino-9H-purin-9-yl)-3-hydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]phosphonate
[Na+].[Na+].NC1=C2N=CN(C3CC(O)C(COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)O3)C2=NC=N1
9000-30-0
C10H14N5Na2O12P3
535.15
JEKDCIBJADJZSK-UHFFFAOYNA-L
disodium hydrogen [({[5-(6-amino-9H-purin-9-yl)-3-hydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]phosphonate
[Na+].[Na+].NC1=C2N=CN(C3CC(O)C(COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)O3)C2=NC=N1

GU110, 9000-30-0

Spectrum™ Guar Gum, FCC is an economical thickening agent because it has almost eight times the water-thickening potency of cornstarch. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Ash 0.015
Percent Purity 70%
Lead (Pb) 2 mg/kg
Loss on Drying 0.15
Packaging Poly Pail
Quantity 2.5 kg