Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Vis-Tech™ Y9294 Polyurethane-Coated Cut-Resistant Gloves

Catalog Number 19048788
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Vis-Tech™ Y9294 Polyurethane-Coated Cut-Resistant Gloves are constructed with a unique blend of high performance fibers and stainless steel to provide comfort and cut resistance. Polyurethane coating on the palm enhances grip capabilities when handling small parts.

  • Constructed from a blend of high performance fibers and stainless steel
  • 13-gauge shell provides ANSI Level A4 cut protection
  • Polyurethane coating on the palm enhances grip and tactility
  • Hi-vis coloring increases visibility during low light applications
  • Open, uncoated back circulates air to keep hands cool and comfortable
ANSI Abrasion Rating ANSI 5
Certifications/Compliance ASTM F1790-97, ASTM F2992-15, ASTM D3889-10
Coating Material Polyurethane
Color Hi-vis Orange, Black
Cut Resistance Rating ANSI A4
Hand Specific
Material HPPE, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size X-Large
EN388 Standard EN 388:4543
For Use With (Application) Assembly, construction, metal fabrication
Glove Cuff Style Knit
Model Y9294XL
Series FlexTech™ Vis-Tech Y9294
Yarn Gauge 13
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit