Guardian Rainwear Coveralls

Manufacturer: Guardian Protective Wear 756Y3XL

 View more versions of this product

Catalog No. 19-812-160

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance NFPA 701
    Material Vinyl
    Seam Type Sewn and tape sealed
    Color Yellow
    Size 3X-Large
    For Use With (Application) Splash protection from most chemicals
    Size (English) Waist 33.5 in.
    Size (Metric) Waist 85.09cm
    Includes Adjustable drawstring waist, plain front

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit