Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GUCA2A Rabbit anti-Human, Polyclonal, liquid, OriGene

Rabbit Polyclonal Antibody
Supplier: OriGene TA890064

Description
Endogenous activator of intestinal guanylate cyclase. It stimulates this enzyme through the same receptor binding region as the heat-stable enterotoxins.Specifications
GUCA2A | |
Polyclonal | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q02747 | |
GUCA2A | |
Recombit protein of human GUCA2A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
GUCA2A | |
GCAP-I, GUCA2, STARA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2980 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title