Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GUCY1B3 Polyclonal Antibody
GUCY1B3 Polyclonal Antibody
Supplier: Thermo Scientific PA5109799
Description
GUCY1B3 Polyclonal Antibody for Western Blot, IHC

Specifications
GUCY1B3 | |
Polyclonal | |
Unconjugated | |
GUCY1B1 | |
beta 1 sGC; GCbeta1; GCS beta1; GCS beta3; GC-SB3; GCSbeta1; GC-S-beta-1; GCS-beta1; GCS-beta-1; GCSbeta3; GCS-beta3; GCS-beta-3; guanylate cyclase 1 soluble subunit beta; guanylate cyclase 1 soluble subunit beta 1; guanylate cyclase 1, soluble, beta 3; guanylate cyclase 70 kDa subunit; guanylate cyclase soluble subunit beta-1; Guanylate cyclase soluble subunit beta-3; guanylate cyclase, soluble, beta 1; GUC1B3; GUCB3; GUCSB3; Gucy1b1; Gucy1b3; nitric oxide-sensitive guanylyl cyclase beta 1 subunit; NO-sensitive GC beta 1 subunit; NO-sensitive guanylyl cyclase beta 1 subunit; SGC; sgc1b1; si:ch73-34b5.3; soluble guanylate cyclase 1 beta 3; soluble guanylate cyclase beta-1 subunit; soluble guanylate cyclase small subunit; soluble guanylate cyclase small subunit beta 1; soluble guanylyl cyclase beta1 subunit | |
Rabbit | |
Affinity Chromatography | |
RUO | |
25202, 2983, 54195 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.08 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O54865, P20595, Q02153 | |
GUCY1B1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human Guanylyl Cyclase beta 1 (Guanylyl Cyclase beta 1 (GUCY1B3)) (NP_0008481). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction