Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GUF1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | GUF1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17968320
|
Novus Biologicals
NBP17968320UL |
20 μL |
Each for $152.22
|
|
NBP179683
|
Novus Biologicals
NBP179683 |
100 μL |
Each for $436.00
|
|
Description
GUF1 Polyclonal specifically detects GUF1 in Mouse samples. It is validated for Western Blot.Specifications
GUF1 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
EC 3.6.5.-, EF-4, Elongation factor 4 homolog, FLJ13220, GTPase GUF1, GTPase of unknown function 1, GTP-binding protein GUF1 homolog, GUF1 GTPase homolog (S. cerevisiae), Ribosomal back-translocase, translation factor GUF1, mitochondrial | |
GUF1 | |
Primary | |
72 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_766299 | |
60558 | |
IgG | |
Affinity Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title