Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Guinea Pig IgG ELISA Kit (Colorimetric)

CLU Colorimetric ELISA Kit for study of Apolipoproteins

$502.00

Specifications

Sensitivity 62.5μg/mL
Sample Type Plasma, Serum
Research Discipline Immunology
Synonym Immunoglobulin heavy constant gamma 1 (G1m marker)
Storage Requirements Store unopened product at 2° to 8°C Protect from light. Do not use past expiration date.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB600661KT View Documents Novus Biologicals™
NBP2600661KIT
1 Kit Each for $502.00
Add to cart
 
Description

Description

The Guinea Pig IgG ELISA Kit (Colorimetric) measures guinea pig IgG in serum, plasma.
Specifications

Specifications

62.5μg/mL
Immunology
Store unopened product at 2° to 8°C Protect from light. Do not use past expiration date.
ELISA
62.5 to 1000μg/mL
Plasma, Serum
Immunoglobulin heavy constant gamma 1 (G1m marker)
IgG
ELISA Kit (Colorimetric)
50 to 100 μL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit