Learn More
Guinea Pig IgG, F(ab), Biotin, Isotype control, Novus Biologicals™
Guinea Pig Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197106
Catalog No. NBP197106
Add to cart
Description
Description
IgG F(ab) Isotype Control antibody specifically detects IgG F(ab) in Guinea Pig samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
IgG F(ab)
Biotin
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Sodium Azide
Guinea Pig
Ion exchange chromatography
RUO
Primary
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
LYOPH
Restore with 1.0mL of deionized water (or equivalent).
Conjugated
IgG2
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Guinea Pig IgG, anti-Guinea Pig IgG F(ab)2 and anti-Guinea Pig Serum. No reaction was observed against anti-Guinea Pig IgG F(c) or anti-Papain.
Guinea Pig