Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GVS ABLUO™ 25mm Syringe Filters, Non-sterile
Quick and efficient filtration of samples and all kind of solutions, solvents or gases
Supplier: GVS FJ25BNPPT002AD01
Description
- Lower hold-up volume
- Increased operating pressure up to 75 psi
- Each filter is labeled with the specific filter material and pore size for easy identification
- Housing injected in Polypropylene or Acrylic
- Membrane Material: Cellulose Acetate, Nylon, PES, Nitrocellulose, Regenerated Cellulose, PVDF, PTFE and PE, Glass Fiber
- Membrane Diameter: 25mm and Housing Diameter: 18mm
- Multifunctional Syringe Filters: equipped with luer-lock or luer-slip male connections for different applications
Applications:
- HPLC sample preparation
- Biological fluids
- Buffer solutions
- Sterile filtering of tissue culture media
- Protein aqueous solutions
Specifications
Transparent | |
0.20 μm | |
4.6 cm2 | |
Female luer lock | |
500/Pk. |
PTFE | |
Non-sterile | |
Polypropylene | |
Male luer lock | |
25 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction