Learn More
GVS Maine FAST™ PAK Protein Array Kits
Manufacturer:  GVS Maine 10485319
 View more versions of this product
Catalog No. 09-977-718
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Protein Array kit
ELISA format (sandwich assay) experiments using antibody arrays, Reverse phase protein arrays using cell or tissue lysates, Purified protein arrays, Antigen arrays for antibody screening and autoimmune detection, Carbohydrate arrays, Lipids and other materials which can be arrayed on nitrocellulose
10 FAST Slides, 10 incubation chambers, 10 ml Protein Arraying Buffer (2X), 15 ml Protein Array Blocking Buffer, and 125 ml Protein Array Washing Buffer (10X) and 1 array pad