Specifications
Protein Array Kit | |
ELISA format (sandwich assay) experiments using antibody arrays, Reverse phase protein arrays using cell or tissue lysates, Purified protein arrays, Antigen arrays for antibody screening and autoimmune detection, Carbohydrate arrays, Lipids and other materials which can be arrayed on nitrocellulose | |
10 FAST Slides, 10 incubation chambers, 10 ml Protein Arraying Buffer (2X), 15 ml Protein Array Blocking Buffer, and 125 ml Protein Array Washing Buffer (10X) and 8 array pads |