GVS Maine FAST™ PAK Protein Array Kits
Manufacturer: GVS Maine 10485325
 View more versions of this product
Catalog No. 09-977-720
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) ELISA format (sandwich assay) experiments using antibody arrays, Reverse phase protein arrays using cell or tissue lysates, Purified protein arrays, Antigen arrays for antibody screening and autoimmune detection, Carbohydrate arrays, Lipids and other materials which can be arrayed on nitrocellulose
    Includes 10 FAST Slides, 10 incubation chambers, 10 ml Protein Arraying Buffer (2X), 15 ml Protein Array Blocking Buffer, and 125 ml Protein Array Washing Buffer (10X) and 16 array pads
    Type Protein Array kit
  • Description & Specifications
Related Products
Product Content Feedback