Learn More
GVS Nitrocellulose Hybridization and Transfer Membranes: Rolls
Require vacuum baking to fix DNA
$513.50 - $1058.00
Catalog Number | Mfr. No. | Length (Metric) | Material | Pore Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Material | Pore Size | Price | Quantity & Availability | |||
EP2HY00010
|
GVS
1215458 |
30 cm | NitroBind | 0.22 μm |
Each for $1,038.00
|
|
|||
WP2HY00010
|
GVS
1212632 |
30 cm | NitroPure | 0.22 μm |
Each for $1,058.00
|
|
|||
EP2HYB0010
|
GVS
1215470 |
15 cm | NitroBind | 0.22 μm |
Each for $514.00
|
|
|||
EP2HYA0010
|
GVS
1215469 |
20 cm | NitroBind | 0.22 μm |
Each for $705.00
|
|
|||
EP4HYA0010
|
GVS
1215483 |
20 cm | NitroBind | 0.45 μm |
Each for $650.00
|
|
|||
EP4HYB0010
|
GVS
1215484 |
15 cm | NitroBind | 0.45 μm |
Each for $513.50
|
|
|||
WP2HYA0010
|
GVS
1212690 |
20 cm | NitroPure | 0.22 μm |
Each for $575.00
|
|
|||
WP4HY00010
|
GVS
1212590 |
30 cm | NitroPure | 0.45 μm |
Each for $937.50
|
|
|||
WP4HYA0010
|
GVS
1212602 |
20 cm | NitroPure | 0.45 μm |
Each for $601.50
|
|
|||
EP4HY00010
|
GVS
1215471 |
30 cm | NitroBind | 0.45 μm |
Each for $941.50
|
|
|||
Description
Nitrocellulose Unsupported Membrane (EP)
- Highest sensitivity for protein blotting and immunoblotting, or other procedures requiring optimum resolution
- Easily blocked
- Wets out naturally and does not require methanol like PVDF membranes
- High sensitivity but with added strength of a polyester matrix
- Surfactant-free membranes have no additives to interfere with binding capacity
Specifications
3 m | |
Roll |