Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ GW 1929 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Description
Highly selective orally active peroxisome proliferator-activated receptor (PPAR)γ agonist (pEC50 values are 8.05, < 4 and < 4 for human PPARγ, PPARα and PPARδ receptors respectively). Decreases glucose, fatty acid and triglyceride levels following oral administration in vivo.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 532.03 |
Purity | >98% |
Molecular Weight (g/mol) | 532.037 |
Chemical Name or Material | GW 1929 hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction