Tocris™ GW 438014A

NPY Y5 receptor antagonist

Manufacturer: Tocris™ 4669/50

 View more versions of this product

Catalog No. 46-695-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors GW 438014A
    Purity >99
    Molecular Weight 437.51
    Quantity 50mg
    Product Type GW 438014A

    Neuropeptide (NPY) Y5 receptor antagonist (IC50 = 210nM). Reduces NPY-induced food intake in rodents and decreases weight gain in obese rats.

    • Chemical Name: N-[1-(2-Phenylethyl)-1H-benzimidazol-2-yl]benzamide methanesulfonate
    • Formula: C22H19N3O.CH3SO3H
    • CAS Number: 469861-49-2
    • Solubility: Soluble to 100mM in DMSO and to 50mM in ethanol
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit