Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GW 441756, Tocris Bioscience™

Catalog Number p-4784007
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
504433-23-2
C17H13N3O
275.311
NXNQLECPAXXYTR-UKTHLTGXSA-N
1,3-dihydro-3-1-methyl-1h-indol-3-yl methylene-2h-pyrrolo 3,2-b pyridin-2-one, 3e-3-1-methylindol-3-yl methylidene-1h-pyrrolo 3,2-b pyridin-2-one, e-3-1-methyl-1h-indol-3-yl methylene-1h-pyrrolo 3,2-b pyridin-2 3h-one, 3-e-1-methyl-1h-indole-3-yl methylene-2,3-dihydro-1h-pyrrolo 3,2-b pyridine-2-one
22228376
(3E)-3-[(1-methylindol-3-yl)methylidene]-1H-pyrrolo[3,2-b]pyridin-2-one
CN1C=C(C2=CC=CC=C21)C=C3C4=C(C=CC=N4)NC3=O
This item is not returnable. View return policy
504433-23-2
C17H13N3O
275.311
NXNQLECPAXXYTR-UKTHLTGXSA-N
1,3-dihydro-3-1-methyl-1h-indol-3-yl methylene-2h-pyrrolo 3,2-b pyridin-2-one, 3e-3-1-methylindol-3-yl methylidene-1h-pyrrolo 3,2-b pyridin-2-one, e-3-1-methyl-1h-indol-3-yl methylene-1h-pyrrolo 3,2-b pyridin-2 3h-one, 3-e-1-methyl-1h-indole-3-yl methylene-2,3-dihydro-1h-pyrrolo 3,2-b pyridine-2-one
22228376
(3E)-3-[(1-methylindol-3-yl)methylidene]-1H-pyrrolo[3,2-b]pyridin-2-one
CN1C=C(C2=CC=CC=C21)C=C3C4=C(C=CC=N4)NC3=O

Potent, selective TrkA inhibitor

Potent, selective inhibitor of the NGF receptor tyrosine kinase A (TrkA) (IC50 = 2 nM). Displays > 100-fold selectivity over a range of other kinases.
Quantity 10 mg
Formula Weight 275.31
Percent Purity >99%
Chemical Name or Material GW 441756
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit