Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GW 627368, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective prostanoid EP4 receptor competitive antagonist with additional affinity at TP receptors (pKi values are 7.0 and 6.8 in competition radioligand bioassays). Affinity for all other prostanoid receptors is < 5.3. Inhibits U-46619 induced human platelet aggregation.
Specifications
Quantity | 10 mg |
Formula Weight | 544.62 |
Percent Purity | >98% |
Chemical Name or Material | GW 627368 |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction