Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GW 791343 hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
309712-55-8
C20H27Cl3F2N4O
483.81
WSBRAHWNJBXXJM-UHFFFAOYSA-N
gw 791343 hydrochloride, gw791343 trihydrochloride, 2-3,4-difluorophenyl amino-n-2-methyl-5-1-piperazinylmethyl phenyl-acetamide trihydrochloride, 2-3,4-difluorophenyl amino-n-2-methyl-5-piperazin-1-ylmethyl phenyl acetamide trihydrochloride, d05exn, gw-791343 hydrochloride, gw 791343 trihydrochloride, c20h24f2n4o.3clh, gw 791343 trihydrochloride;gw-791343 trihydrochloride
9848159
2-(3,4-difluoroanilino)-N-[2-methyl-5-(piperazin-1-ylmethyl)phenyl]acetamide;trihydrochloride
CC1=C(C=C(C=C1)CN2CCNCC2)NC(=O)CNC3=CC(=C(C=C3)F)F.Cl.Cl.Cl
This item is not returnable. View return policy
309712-55-8
C20H27Cl3F2N4O
483.81
WSBRAHWNJBXXJM-UHFFFAOYSA-N
gw 791343 hydrochloride, gw791343 trihydrochloride, 2-3,4-difluorophenyl amino-n-2-methyl-5-1-piperazinylmethyl phenyl-acetamide trihydrochloride, 2-3,4-difluorophenyl amino-n-2-methyl-5-piperazin-1-ylmethyl phenyl acetamide trihydrochloride, d05exn, gw-791343 hydrochloride, gw 791343 trihydrochloride, c20h24f2n4o.3clh, gw 791343 trihydrochloride;gw-791343 trihydrochloride
9848159
2-(3,4-difluoroanilino)-N-[2-methyl-5-(piperazin-1-ylmethyl)phenyl]acetamide;trihydrochloride
CC1=C(C=C(C=C1)CN2CCNCC2)NC(=O)CNC3=CC(=C(C=C3)F)F.Cl.Cl.Cl

P2X7 allosteric modulator

P2X7 allosteric modulator. Exhibits species-specific activity; acts as a negative allosteric modulator of human P2X7 (pIC50 = 6.9 - 7.2) and a positive allosteric modulator of rat P2X7.
Quantity 50mg
Formula Weight 483.81
Percent Purity >99%
Chemical Name or Material GW 791343 hydrochloride

Recommended Storage : Desiccate at Room Temperature