Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ GW 9662

Click to view available options
Quantity:
10mg
50mg
22978-25-2
C13H9ClN2O3
276.676
MFCD01215270
DNTSIBUQMRRYIU-UHFFFAOYSA-N
2-chloro-5-nitrobenzanilide,2-chloro-5-nitro-n-phenyl-benzamide,2-chloro-5-nitro-n-4-phenylbenzamide,2-chloro-5-nitrophenyl-n-benzamide,tocris-1508,spectrum5_001989,lopac-m-6191,d00acx,d05thb,dsstox_cid_20723
644213
CHEBI:79993
2-chloro-5-nitro-N-phenylbenzamide
C1=CC=C(C=C1)NC(=O)C2=C(C=CC(=C2)[N+](=O)[O-])Cl
22978-25-2
C13H9ClN2O3
276.676
MFCD01215270
DNTSIBUQMRRYIU-UHFFFAOYSA-N
2-chloro-5-nitrobenzanilide,2-chloro-5-nitro-n-phenyl-benzamide,2-chloro-5-nitro-n-4-phenylbenzamide,2-chloro-5-nitrophenyl-n-benzamide,tocris-1508,spectrum5_001989,lopac-m-6191,d00acx,d05thb,dsstox_cid_20723
644213
CHEBI:79993
2-chloro-5-nitro-N-phenylbenzamide
C1=CC=C(C=C1)NC(=O)C2=C(C=CC(=C2)[N+](=O)[O-])Cl

An irreversible PPAR→ antagonist

Quantity 50mg
Color White
Formula Weight 276.68
Physical Form Powder
Chemical Name or Material GW 9662

P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P363-P501c

H317-H319

TSCA : No

Recommended Storage : Ambient temperatures