Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GW 9662, Tocris Bioscience™

Catalog Number p-4783504
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
22978-25-2
C13H9ClN2O3
276.676
DNTSIBUQMRRYIU-UHFFFAOYSA-N
2-chloro-5-nitrobenzanilide, 2-chloro-5-nitro-n-phenyl-benzamide, 2-chloro-5-nitro-n-4-phenylbenzamide, 2-chloro-5-nitrophenyl-n-benzamide, tocris-1508, spectrum5_001989, lopac-m-6191, d00acx, d05thb, dsstox_cid_20723
644213
CHEBI:79993
2-chloro-5-nitro-N-phenylbenzamide
C1=CC=C(C=C1)NC(=O)C2=C(C=CC(=C2)[N+](=O)[O-])Cl
This item is not returnable. View return policy
22978-25-2
C13H9ClN2O3
276.676
DNTSIBUQMRRYIU-UHFFFAOYSA-N
2-chloro-5-nitrobenzanilide, 2-chloro-5-nitro-n-phenyl-benzamide, 2-chloro-5-nitro-n-4-phenylbenzamide, 2-chloro-5-nitrophenyl-n-benzamide, tocris-1508, spectrum5_001989, lopac-m-6191, d00acx, d05thb, dsstox_cid_20723
644213
CHEBI:79993
2-chloro-5-nitro-N-phenylbenzamide
C1=CC=C(C=C1)NC(=O)C2=C(C=CC(=C2)[N+](=O)[O-])Cl

Selective covalent PPARγ antagonist

GW 9662 is a selective irreversible PPARγ antagonist (IC50 values are 3.3, 32 and 2000 nM for PPARγ, PPARα and PPARδ respectively). Blocks the inhibition of osteoclast formation induced by IL-4 in the low micromolar range (1-2 μM), therefore is more potent than BADGE. Anticancer, inhibits growth of human mammary tumor cell lines.
Quantity 10 mg
Formula Weight 276.68
Percent Purity >98%
Chemical Name or Material GW 9662
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit